Anti-JAM3 Mouse Monoclonal Antibody [clone: 1D3]
Catalog # ABNOH00083700-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Junctional Adhesion Molecule 3
- Antigen symbol:JAM3
- Clonality:Monoclonal
- Clone:1D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:83700
- Antigen synonyms:JAM-3|JAM-C|JAMC|JAM-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: JAM3
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human