Anti-FBXO11 Mouse Polyclonal Antibody
ABNOH00080204-B01P
: Abnova
- Antibody type:Primary
- Antigen name:F-box protein 11
- Antigen symbol:FBXO11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80204
- Antigen synonyms:UG063H01|UBR6|PRMT9|VIT1|FBX11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: FBXO11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human