Anti-IRX3 Mouse Monoclonal Antibody [clone: 3D8]
Catalog # ABNOH00079191-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Iroquois Homeobox Protein 3
- Antigen symbol:IRX3
- Clonality:Monoclonal
- Clone:3D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79191
- Antigen synonyms:Homeodomain protein IRXB1|Iroquois-class homeodomain protein IRX-3|Iroquois homeobox protein 3|IRXB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IRX3
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human