Anti-ACBD3 Mouse Monoclonal Antibody [clone: 2G2]
ABNOH00064746-M01
: Abnova
- Antibody type:Primary
- Antigen name:Acyl-CoA Binding Domain Containing 3
- Antigen symbol:ACBD3
- Clonality:Monoclonal
- Clone:2G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:64746
- Antigen synonyms:GOCAP 1|Peripherial benzodiazepine receptor associated protein|PAP 7|ACBD 3|Golgi complex associated protein 1 60kDa|GCP60|Golgi phosphoprotein 1|GCP 60|Golgi complex-associated protein 1|Acyl Coenzyme A binding domain containing 3|GOLPH 1|GOCAP1|Acyl CoA binding domain containing protein 3|Peripheral benzodiazepine receptor associated protein PAP7|PKA RIalpha associated protein.|PBR associated protein|ACBD3|PAP7|Golgi complex associated protein 1|GCP60_HUMAN|PBR and PKA associated protein 7|Golgi resident protein GCP60|PBR- and PKA-associated protein 7|Acyl-CoA-binding domain-containing protein 3|Peripheral benzodiazepine receptor-associated protein PAP7|GOLPH1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ACBD3
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat