Anti-USP31 Mouse Monoclonal Antibody [clone: 3B6]
ABNOH00057478-M01
: Abnova
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 31
- Antigen symbol:USP31
- Clonality:Monoclonal
- Clone:3B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57478
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: USP31
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human