Anti-MYOZ2 Mouse Monoclonal Antibody [clone: 1D4]
Catalog # ABNOH00051778-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Myozenin 2
- Antigen symbol:MYOZ2
- Clonality:Monoclonal
- Clone:1D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51778
- Antigen synonyms:C4orf5|CMH16|CS-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MYOZ2
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human