Anti-MED31 Mouse Monoclonal Antibody [clone: 2C8]
ABNOH00051003-M01A
: Abnova
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 31
- Antigen symbol:MED31
- Clonality:Monoclonal
- Clone:2C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51003
- Antigen synonyms:Soh1|CGI-125|3110004H13Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: MED31
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human