Anti-GOLIM4 Mouse Monoclonal Antibody [clone: 5E11]
ABNOH00027333-M01
: Abnova
- Antibody type:Primary
- Antigen name:Golgi integral membrane protein 4
- Antigen symbol:GOLIM4
- Clonality:Monoclonal
- Clone:5E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:27333
- Antigen synonyms:GIMPC|GOLPH4|P138|GPP130
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GOLIM4
Clonality: Monoclonal
Clone: 5E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human