Anti-DFNB31 Mouse Monoclonal Antibody [clone: 2D12]
Catalog # ABNOH00025861-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:deafness, autosomal recessive 31
- Antigen symbol:DFNB31
- Clonality:Monoclonal
- Clone:2D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:25861
- Antigen synonyms:WI|PDZD7B|WHRN|CIP98|USH2D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DFNB31
Clonality: Monoclonal
Clone: 2D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human