Anti-EID1 Mouse Polyclonal Antibody
ABNOH00023741-B01P
: Abnova
- Antibody type:Primary
- Antigen name:EP300 interacting inhibitor of differentiation 1
- Antigen symbol:EID1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23741
- Antigen synonyms:PNAS-22|IRO45620|PTD014|EID-1|RBP21|C15orf3|CRI1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: EID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human