Anti-DNPEP Mouse Monoclonal Antibody [clone: 2F9-3A7]
Catalog # ABNOH00023549-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:aspartyl aminopeptidase
- Antigen symbol:DNPEP
- Clonality:Monoclonal
- Clone:2F9-3A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23549
- Antigen synonyms:ASPEP, DAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DNPEP
Clonality: Monoclonal
Clone: 2F9-3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human