Anti-ISCU Mouse Monoclonal Antibody [clone: 3D12-1D10]
Catalog # ABNOH00023479-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Iron-Sulfur Cluster Assembly Enzyme
- Antigen symbol:ISCU
- Clonality:Monoclonal
- Clone:3D12-1D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23479
- Antigen synonyms:NIFUN|NIFU|2310020H20Rik|hnifU|HML|ISU2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ISCU
Clonality: Monoclonal
Clone: 3D12-1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human