Anti-LILRA3 Mouse Monoclonal Antibody [clone: 2E9]
Catalog # ABNOH00011026-M01
Supplier: Abnova
Specifications
- Antistofftype:Primary
- Antigen-navn:Leukocyte immunoglobulin-like Receptor subfamily A (with TM Domain) member 3
- Antigensymbol:LILRA3
- Klonalitet:Monoclonal
- Klon:2E9
- Konjugering:Unconjugated
- ELISA:Yes
- Vert:Mouse
- Reaktivitet:Human
- Western blot:Yes
- Kyssabsorpsjon:No
- Gen-ID:11026
- Antigen synonymer:LIR4|CD85E|ILT-6|LIR-4|ILT6|HM31|HM43
- Lagringstemp:Cool
- Frakttemp.:Ice
- Str:100 µg
- Fp:100 µg
Specifications
About this item
Type: Primary
Antigen: LILRA3
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human