Anti-NEK6 Mouse Monoclonal Antibody [clone: 3B5]
ABNOH00010783-M01
: Abnova
- Antibody type:Primary
- Antigen name:NIMA-related kinase 6
- Antigen symbol:NEK6
- Clonality:Monoclonal
- Clone:3B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:10783
- Antigen synonyms:SID|SID6-1512|RP11-101K10.6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NEK6
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human