Order Entry
Norway
ContactUsLinkComponent
Anti-P3H4 Rabbit Polyclonal Antibody
Anti-P3H4 Rabbit Polyclonal Antibody
  ABNOH00010609-D01
 :  Abnova
undefined
Anti-P3H4 Rabbit Polyclonal Antibody
  ABNOH00010609-D01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Prolyl 3-Hydroxylase 4
  • Antigen symbol:
    P3H4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    10609
  • Antigen synonyms:
    SC65|LEPREL4|NO55|NOL55
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: P3H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human