Anti-MED27 Mouse Monoclonal Antibody [clone: 8B8]
Catalog # ABNOH00009442-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 27
- Antigen symbol:MED27
- Clonality:Monoclonal
- Clone:8B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9442
- Antigen synonyms:CRSP8|CRSP34|TRAP37|CRAP34
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: MED27
Clonality: Monoclonal
Clone: 8B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human