Anti-PSMG1 Mouse Monoclonal Antibody [clone: 4F2]
ABNOH00008624-M01
: Abnova
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) Assembly Chaperone 1
- Antigen symbol:PSMG1
- Clonality:Monoclonal
- Clone:4F2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:8624
- Antigen synonyms:PAC-1|PAC1|C21LRP|LRPC21|DSCR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PSMG1
Clonality: Monoclonal
Clone: 4F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human