Order Entry
Norway
ContactUsLinkComponent
 
Anti-STK16 Mouse Monoclonal Antibody [clone: M2]
  ABNOH00008576-M02
 :  Abnova
undefined
Anti-STK16 Mouse Monoclonal Antibody [clone: M2]
  ABNOH00008576-M02
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Serine/threonine Kinase 16
  • Antigen symbol:
    STK16
  • Clonality:
    Monoclonal
  • Clone:
    M2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    8576
  • Antigen synonyms:
    hPSK|Myristoylated and palmitoylated serine-threonine kinase|Protein kinase PKL12|TSF-1|MPSK|TGF-beta stimulated factor 1|Serine/threonine protein kinase 16
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: STK16
Clonality: Monoclonal
Clone: M2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human