Order Entry
Norway
ContactUsLinkComponent
Anti-VCP Mouse Monoclonal Antibody [clone: 2B2]
Anti-VCP Mouse Monoclonal Antibody [clone: 2B2]
Catalog # ABNOH00007415-M18
Supplier:  Abnova
undefined
Anti-VCP Mouse Monoclonal Antibody [clone: 2B2]
Catalog # ABNOH00007415-M18
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Valosin containing protein
  • Antigen symbol:
    VCP
  • Clonality:
    Monoclonal
  • Clone:
    2B2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    7415
  • Antigen synonyms:
    ALS14|HEL-S-70|IBMPFD1|TERA|p97|HEL-220|IBMPFD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: VCP
Clonality: Monoclonal
Clone: 2B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human