Anti-STAT5B Mouse Monoclonal Antibody [clone: 1C2]
ABNOH00006777-M04
: Abnova
- Antistofftype:Primary
- Antigen-navn:Signal transducer and activator of transcription 5B
- Antigensymbol:STAT5B
- Klonalitet:Monoclonal
- Klon:1C2
- Konjugering:Unconjugated
- ELISA:Yes
- Vert:Mouse
- Reaktivitet:Human
- Western blot:Yes
- Kyssabsorpsjon:No
- Gen-ID:6777
- Antigen synonymer:STAT5|Signal transducer and activator of transcription 5B|Transcription factor STAT5B|Stat5b.|STA5B_HUMAN
- Lagringstemp:Cool
- Frakttemp.:Ice
- Str:100 µg
- Fp:100 µg
Type: Primary
Antigen: STAT5B
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human