Anti-PYGM Mouse Monoclonal Antibody [clone: 3D8]
Catalog # ABNOH00005837-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phosphorylase, glycogen, muscle
- Antigen symbol:PYGM
- Clonality:Monoclonal
- Clone:3D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5837
- Antigen synonyms:PYGM|muscle form|Glycogen phosphorylase|Myophosphorylase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PYGM
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human