Anti-SMAD4 Mouse Monoclonal Antibody [clone: 3B9]
Catalog # ABNOH00004089-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:SMAD family member 4
- Clonality:Monoclonal
- Clone:3B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4089
- Antigen synonyms:Drosophila|mothers against decapentaplegic homolog 4,SMAD 4|hSMAD4|SMAD|mothers against decapentaplegic homolog 4|mothers against DPP homolog 4|MAD|deleted in pancreatic carcinoma locus 4|4|MAD homolog 4|mothers against decapentaplegic|deletion target in pancreatic carcinoma 4|homolog of
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: SMAD4
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human