Anti-LIMK1 Mouse Monoclonal Antibody [clone: 2E9]
Catalog # ABNOH00003984-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:LIM Domain Kinase 1
- Antigen symbol:LIMK1
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3984
- Antigen synonyms:LIMK-1|KIZ-1|LIM domain kinase 1|EC 2.7.11.1|LIMK|kinase LIMK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LIMK1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human