Anti-IgD Mouse Polyclonal Antibody
ABNOH00003495-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Immunoglobulin D
- Antigen symbol:IgD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3495
- Antigen synonyms:Igh-5|IGHD
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: IgD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human