Anti-GRM5 Mouse Monoclonal Antibody [clone: 1B3]
ABNOH00002915-M01
: Abnova
- Antibody type:Primary
- Antigen name:Glutamate receptor, metabotropic 5
- Antigen symbol:GRM5
- Clonality:Monoclonal
- Clone:1B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2915
- Antigen synonyms:GPRC1E|MGLUR5|GRM5|Metabotropic glutamate receptor 5 [Precursor]|mGluR5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GRM5
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human