Anti-FLT3L Mouse Monoclonal Antibody [clone: 4C4]
ABNOH00002323-M07
: Abnova
- Antibody type:Primary
- Antigen name:Fms-related tyrosine kinase 3 ligand
- Antigen symbol:FLT3LG
- Clonality:Monoclonal
- Clone:4C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2323
- Antigen synonyms:FLT3L|Fms-related tyrosine kinase 3 ligand|SL cytokine|FLT3LG|FL|Flt3 ligand
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: FLT3L
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human