Order Entry
Norway
ContactUsLinkComponent
 
Anti-FOXC2 Mouse Monoclonal Antibody [clone: 2H3]
  ABNOH00002303-M02A
 :  Abnova
undefined
Anti-FOXC2 Mouse Monoclonal Antibody [clone: 2H3]
  ABNOH00002303-M02A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Forkhead Box C2
  • Antigen symbol:
    FOXC2
  • Clonality:
    Monoclonal
  • Clone:
    2H3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    2303
  • Antigen synonyms:
    MFH1|MFH-1|FKHL14|LD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: FOXC2
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human