Anti-BDKRB2 Mouse Monoclonal Antibody [clone: 3F6]
Catalog # ABNOH00000624-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Bradykinin Receptor B2
- Antigen symbol:BDKRB2
- Clonality:Monoclonal
- Clone:3F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:624
- Antigen synonyms:BKR2|BK2|BRB2|BK-2|B2R
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BDKRB2
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human