Anti-AMELX Mouse Polyclonal Antibody
ABNOH00000265-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Amelogenin X-linked
- Antigen symbol:AMELX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:265
- Antigen synonyms:AI1E|AMG|AMGX|AMGL|ALGN|AIH1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: AMELX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human