Anti-ALPPL2 Mouse Monoclonal Antibody [clone: 2B3]
Catalog # ABNOH00000251-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase, Placental-Like 2
- Antigen symbol:ALPPL2
- Clonality:Monoclonal
- Clone:2B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:251
- Antigen synonyms:ALPPL|ALPG|GCAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ALPPL2
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human