Anti-PGM3 Mouse Monoclonal Antibody [clone: OTI1A11]
Catalog # ORIGTA504859
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:phosphoglucomutase 3
- Antigen symbol:PGM3
- Clonality:Monoclonal
- Clone:OTI1A11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:59.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: PGM3 (phosphoglucomutase 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: