Order Entry
Norway
ContactUsLinkComponent
Anti-CBWD1 Mouse Monoclonal Antibody [clone: OTI4D4]
Anti-CBWD1 Mouse Monoclonal Antibody [clone: OTI4D4]
Catalog # ORIGTA501609
Supplier:  OriGene
undefined
Anti-CBWD1 Mouse Monoclonal Antibody [clone: OTI4D4]
Catalog # ORIGTA501609
Supplier:  OriGene

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    COBW domain containing 1
  • Antigensymbol:
    CBWD1
  • Klonalitet:
    Monoclonal
  • Klon:
    OTI4D4
  • Konjugering:
    Unconjugated
  • Vert:
    Mouse
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Kyssabsorpsjon:
    none
  • Skjema:
    Liquid
  • Lagringsbuffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekylvekt:
    43.9 kDa
  • Lagringstemp:
    Cool
  • Frakttemp.:
    Ice
  • Rensing:
    purified by affinity chromatography
  • Fp:
    100 µl

Specifications

About this item

  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CBWD1 (COBW domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: