Order Entry
Norway
ContactUsLinkComponent
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin) [clone: M1/70]
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin) [clone: M1/70]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin) [clone: M1/70]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0112-U100
  • 20-0112-U025
  • Antigen symbol:
  • ITGAM
  • ITGAM
  • Antigen name:
  • Integrin Alpha M
  • Integrin Alpha M
  • Antigen synonyms:
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • M1/70
  • M1/70
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16409 / 3684
  • 16409 / 3684
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information