Anti-TBCD7 Rabbit Polyclonal Antibody
ABGEAP9903C
: Abgent
- Antibody type:Primary
- Antigen name:TBC1 Domain Family Member 7
- Antigen symbol:TBC1D7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:51666
- Antigen synonyms:ASB-4
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:YKAEVNARDDDFKSPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ASB4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 226-255 amino acids from the C-terminal region of human ASB4.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: TBCD7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse