Order Entry
Norway
ContactUsLinkComponent
Anti-Natriuretic Peptide Receptor A (NPR1/ANPA) Rabbit Polyclonal Antibody
Anti-Natriuretic Peptide Receptor A (NPR1/ANPA) Rabbit Polyclonal Antibody
  ABGEAP8111A
 :  Abgent
undefined
Anti-Natriuretic Peptide Receptor A (NPR1/ANPA) Rabbit Polyclonal Antibody
  ABGEAP8111A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Natriuretic peptide A
  • Antigen symbol:
    NPPA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal (1–100 aa)
  • Cross adsorption:
    No
  • Gene ID:
    1475
  • Antigen synonyms:
    PSS4|Cystatin A|AREI|STF1|STFA
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    CGYQVDKNKDDELTGF
  • Storage temperature:
    Cool
  • Concentration:
    0,45 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This CSTA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 69-98 amino acids from the C-terminal region of human CSTA.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: Natriuretic Peptide Receptor A (NPR1/ANPA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat