Order Entry
Norway
ContactUsLinkComponent
Anti-PRKAB1 Rabbit Polyclonal Antibody
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7045A
Supplier:  Abgent
undefined
Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7045A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase, AMP-activated Non-catalytic Subunit Beta 1
  • Antigen symbol:
    PRKAB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal (1–100 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse