Order Entry
Norway
ContactUsLinkComponent
Anti-IL5RA Rabbit Polyclonal Antibody
Anti-IL5RA Rabbit Polyclonal Antibody
Catalog # ABGEAP5235A
Supplier:  Abgent
undefined
Anti-IL5RA Rabbit Polyclonal Antibody
Catalog # ABGEAP5235A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 5 Receptor Alpha
  • Antigen symbol:
    IL5RA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    IL-5R subunit alpha|CDw125|HSIL5R3|IL-5 receptor subunit alpha|interleukin-5 receptor alpha chain|interleukin 5 receptor type 3|IL5R|CD125 antigen|CD125|interleukin-5 receptor subunit alpha
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: IL5RA (interleukin 5 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse