Anti-B2M Rabbit Polyclonal Antibody
Catalog # ABGEAP2771A
Supplier: Abgent
Specifications
- Antistofftype:Primary
- Antigen-navn:Beta-2-Microglobulin
- Antigensymbol:B2M
- Klonalitet:Polyclonal
- Konjugering:Unconjugated
- ELISA:Yes
- Flowcytometri:Yes
- Vert:Rabbit
- ImmunoKjemi:Yes
- Reaktivitet:Human
- Western blot:Yes
- Epitop:N-terminal
- Antigen synonymer:CDABP0092|HDCMA22P|Beta-2-microglobulin|B2M
- Lagringstemp:Cool
- Frakttemp.:Ice
- Fp:400 µl
Specifications
About this item
Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human