Order Entry
Norway
ContactUsLinkComponent
 
Anti-BRF1 Rabbit Polyclonal Antibody
  ABGEAP14801C
 :  Abgent
Anti-BRF1 Rabbit Polyclonal Antibody
  ABGEAP14801C
 :  Abgent

 

  • Antistofftype:
    Primary
  • Antigen-navn:
    BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
  • Antigensymbol:
    BRF1
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Unconjugated
  • ELISA:
    Yes
  • Flowcytometri:
    Yes
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    Center
  • Gen-ID:
    2972
  • Antigen synonymer:
    HEL-S-76p|TFIIIB90|TAF3B2|TAFIII90|GTF3B|hBRF|RF|TF3B90|RNA polymerase III transcription initiation factor 90 kDa subunit|TAF3C|BRF-1|CFDS
  • Lagringsbuffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molekylvekt:
    73.84 kDa
  • Sekvens:
    EIELENSRPKAKGGC
  • Lagringstemp:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Konsentrasjon:
    0,5 mg/ml
  • Frakttemp.:
    4 °C
  • Immunogen:
    This BRF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 314-342 amino acids from the Central region of human BRF1.
  • Rensing:
    Protein A & Peptide affinity purification
  • Fp:
    400 µl

 

 

Type: Primary
Antigen: BRF1 (BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human