Anti-IGSF5 Rabbit Polyclonal Antibody
ABGEAP10623A
: Abgent
- Antibody type:Primary
- Antigen name:Immunoglobulin Superfamily Member 5
- Antigen symbol:IGSF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (1500–1600 aa)
- Cross adsorption:No
- Antigen synonyms:GSF5|IGSF5_HUMAN|Junctional adhesion molecule 4.|Immunoglobulin superfamily 5 like|IgSF5|JAM-4|Immunoglobulin superfamily member 5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: IGSF5 (immunoglobulin superfamily, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human