Order Entry
Norway
ContactUsLinkComponent
Anti-HSPA9 Rabbit Polyclonal Antibody
Anti-HSPA9 Rabbit Polyclonal Antibody
Catalog # ABGEAP10160C
Supplier:  Abgent
Anti-HSPA9 Rabbit Polyclonal Antibody
Catalog # ABGEAP10160C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat Shock 70 kDa Protein 9 (mortalin)
  • Antigen symbol:
    HSPA9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    MOT|GRP-75|CRP40|GRP75|MOT2|HSPA9B|CSA|HEL-S-124m|PBP74|MTHSP75
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: HSPA9 (heat shock 70kDa protein 9 (mortalin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human