Anti-RGL1 Rabbit Polyclonal Antibody
AVIVARP64399_P050
- Antibody type:Primary
- Antigen name:Ral Guanine Nucleotide Dissociation Stimulator-like 1
- Antigen symbol:RGL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23179
- Antigen synonyms:RGL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RGL1 (ral guanine nucleotide dissociation stimulator-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human