Anti-RPL10 Rabbit Polyclonal Antibody
Catalog # AVIVARP64376_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein L10
- Antigen symbol:RPL10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:6134
- Antigen synonyms:DXS648|NOV|DXS648E|L10|AUTSX5|QM
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RPL10 (ribosomal protein L10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: