Anti-MTUS2 Rabbit Polyclonal Antibody
AVIVARP62853_P050
- Antistofftype:Primary
- Antigen-navn:microtubule associated tumor suppressor candidate 2
- Antigensymbol:MTUS2
- Klonalitet:Polyclonal
- Konjugering:Unconjugated
- Vert:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Kyssabsorpsjon:none
- Gen-ID:23281
- Antigen synonymer:CAZIP|KIAA0774|ICIS|TIP150
- Lagringstemp:Cool
- Frakttemp.:Ice
- Fp:50 µG
Type: Primary
Antigen: MTUS2 (microtubule associated tumor suppressor candidate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: