Anti-DDO Rabbit Polyclonal Antibody
Catalog # AVIVARP61287_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:D-aspartate oxidase
- Antigen symbol:DDO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:8528
- Antigen synonyms:DASOX|DDO-2|DDO-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DDO (D-aspartate oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: