Anti-THEG Rabbit Polyclonal Antibody
AVIVARP57810_P050
- Antibody type:Primary
- Antigen name:theg spermatid protein
- Antigen symbol:THEG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:51298
- Antigen synonyms:THEG1|CT56
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: THEG (theg spermatid protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat