Anti-LMF1 Rabbit Polyclonal Antibody
AVIVARP49754_P050
- Antibody type:Primary
- Antigen name:lipase maturation factor 1
- Antigen symbol:LMF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:64788
- Antigen synonyms:TMEM112A|HMFN1876|TMEM112|C16orf26|JFP11
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LMF1 (lipase maturation factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human