Anti-VSIG4 Rabbit Polyclonal Antibody
Catalog # AVIVARP41591_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:V-set and immunoglobulin domain containing 4
- Antigen symbol:VSIG4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:11326
- Antigen synonyms:Z39IG|CRIg
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: VSIG4 (V-set and immunoglobulin domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: