Anti-ZNF677 Rabbit Polyclonal Antibody
AVIVARP36274_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 677
- Antigen symbol:ZNF677
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:342926
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF677 (zinc finger protein 677)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human