Order Entry
Norway
ContactUsLinkComponent
Closed cap, Dino-Lite
Closed cap, Dino-Lite
Catalog # IDCPZT-Z-CC1
Closed cap, Dino-Lite
Catalog # IDCPZT-Z-CC1
Specifications
  • Beskrivelse:
    Closed cap to protect the lens from dust, for polarising AM series (ZT models)
  • Seq:
    03
  • Fp:
    1
Specifications
About this item
Closed cap addition for the polarising AM series (ZT models). Designed to help protect the lens from dust and more.
This snap-on cover fits the AM413ZT, AM4013MZT and AM7013MZT model. Not designed for use with the AD series or AM models without polarisation.